5-Bromo-2-methox-6-methylypyridine 98%

Supplier: Ambeed
Warning

A296623-100G A296623-1G A296623-5G A296623-10G A296623-25G
77002-426EA 135 USD
77002-426 77002-428 77002-430 77881-812 77881-813
5-Bromo-2-methox-6-methylypyridine 98%
5-Bromo-2-methox-6-methylypyridine

Caution: for Research Use Only

Formula: C₇H₈BrNO
MDL Number: MFCD03094943
CAS Number: 126717-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR