5-Iodo-2'-deoxyuridine (Idoxuridine) 98%

Supplier: Ambeed
Warning

Synonyms: 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-iodopyrimidine-2,4(1H,3H)-dione, Idoxuridine, (+)-5-Iodo-2'-deoxyuridine

A133207-100G A133207-1MG A133207-1G A133207-5G A133207-10G
103801-846EA 23 USD
103801-846 103832-770 76972-194 76972-196 76972-198 77298-614 77314-476 77881-056
5-Iodo-2'-deoxyuridine (Idoxuridine) 98%
5-Iodo-2'-deoxyuridine (Idoxuridine)

Caution: for Research Use Only

Formula: C₉H₁₁IN₂O₅
MW: 354.10 g/mol
Melting Pt: 194 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00134656
CAS Number: 54-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR