Phenoxyacetyl chloride ≥98%

Supplier: Ambeed
Danger

A950387-25G
77880-770EA 63 USD
77880-770
Phenoxyacetyl chloride ≥98%
Phenoxyacetyl chloride

Caution: for Research Use Only

Formula: C₈H₇ClO₂
MDL Number: MFCD00000726
CAS Number: 701-99-5
UN: 3265
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR