(S)-1-(Boc-amino)-2-propanol ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
tert-Butyl (S)-(2-hydroxypropyl)carbamate
A918594-1G
A918594-5G
77880-745EA
17
USD
77880-745
77880-746
(S)-1-(Boc-amino)-2-propanol ≥98%
(S)-1-(Boc-amino)-2-propanol
Caution: for Research Use Only
Formula:
C₈H₁₇NO₃ MW: 175.23 g/mol |
MDL Number:
MFCD04974340 CAS Number: 167938-56-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...