Octafluoroadipic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: Perfluoroadipic Acid, Octafluorohexanedioic Acid, Perfluorohexanedioic Acid

A809130-25G
77880-610EA 319 USD
77880-610
Octafluoroadipic acid ≥98%
Octafluoroadipic acid

Caution: for Research Use Only

Formula: C₆H₂F₈O₄
MDL Number: MFCD00066405
CAS Number: 336-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR