Cyanidin-3-O-galactoside chloride ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-3-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)chromenylium chloride

A652650-10MG
77880-372EA 186 USD
77880-372
Cyanidin-3-O-galactoside chloride ≥98%
Cyanidin-3-O-galactoside chloride

Caution: for Research Use Only

Formula: C₂₁H₂₁O₁₁Cl
MW: 484.85 g/mol
MDL Number: MFCD00270387
CAS Number: 27661-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR