D-erythro-Sphingosine 98%

Supplier: Ambeed
Warning

Synonyms: D-Sphingosine, trans-D-erythro-2-Amino-4-octadecene-1,3-diol, (2S,3R,E)-2-Amino-4-octadecene-1,3-diol, 4-Sphingenine, (2S,3R,E)-2-Aminooctadec-4-ene-1,3-diol

A536742-1MG A536742-1G A536742-250MG A536742-100MG A536742-5G A536742-10MG
76695-976EA 8 USD
76695-976 77569-038 77569-040 77572-700 77627-090 77880-164
D-erythro-Sphingosine 98%
D-erythro-Sphingosine

Caution: For research use only.

Formula: C₁₈H₃₇NO₂
MW: 299.50 g/mol
MDL Number: MFCD00036751
CAS Number: 123-78-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR