N-Benzylacetamide 98%

Supplier: Ambeed
Warning

A474806-100MG A474806-1G A474806-25G A474806-5G A474806-100G A474806-500G
76725-820EA 5 USD
76725-820 76725-822 76725-824 76725-826 77669-136 77880-030
N-Benzylacetamide 98%
N-Benzylacetamide

Caution: for Research Use Only

Formula: C₉H₁₁NO
MDL Number: MFCD00059204
CAS Number: 588-46-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR