4-Formyl-3,5-dimethyl-1H-pyrrole-2-carboxylic acid ethyl ester ≥98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 3,5-dimethyl-4-formyl-1H-pyrrole-2-carboxylate

A353945-1G A353945-250MG
77879-762EA 82 USD
77879-762 77879-763
4-Formyl-3,5-dimethyl-1H-pyrrole-2-carboxylic acid ethyl ester ≥98%
4-Formyl-3,5-dimethyl-1H-pyrrole-2-carboxylic acid ethyl ester

Caution: for Research Use Only

Formula: C₁₀H₁₃NO₃
MDL Number: MFCD00030384
CAS Number: 2199-64-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR