Dihydroethidium ≥98%

Supplier: Ambeed
Warning

Synonyms: 3,8-Diamino-5,6-dihydro-5-ethyl-6-phenylphenanthridine, Hydroethidine, DHE, 5-Ethyl-6-phenyl-5,6-dihydrophenanthridine-3,8-diamine, 2,7-Diamino-10-ethyl-9-phenyl-9,10-dihydrophenanthridine

A286650-250MG A286650-50MG A286650-100MG A286650-1G
77626-480EA 462.47 USD
77626-480 77626-482 77626-478 77879-556
Dihydroethidium ≥98%
Dihydroethidium

Caution: for Research Use Only

Formula: C₂₁H₂₁N₃
MDL Number: MFCD00077335
CAS Number: 104821-25-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR