Methyl cholate ≥95%

Supplier: Ambeed
Warning

A223721-1G A223721-25G A223721-100MG A223721-5G
77626-130EA 17.51 USD
77626-130 77626-132 77879-359 77879-360
Methyl cholate ≥95%
Methyl cholate

Caution: for Research Use Only

Formula: C₂₅H₄₂O₅
MDL Number: MFCD00064934
CAS Number: 1448-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR