Methyl-2,3,4-tri-O-benzyl-ɑ-D-glucopyranoside ≥98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2,3,4-Tri-O-benzyl-α-D-glucopyranoside

A167864-1G A167864-5G
77667-342EA 42 USD
77667-342 77879-125
Methyl-2,3,4-tri-O-benzyl-ɑ-D-glucopyranoside ≥98%
Methyl-2,3,4-tri-O-benzyl-alpha-D-glucopyranoside

Caution: for Research Use Only

Formula: C₂₈H₃₂O₆
MDL Number: MFCD06798958
CAS Number: 53008-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR