trans-4-(Boc-amino)cyclohexanemethylamine 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (trans-4-(aminomethyl)cyclohexyl)carbamate

A122855-100MG A122855-10G A122855-250MG A122855-5G A122855-1G A122855-25G
103800-292EA 25 USD
103800-292 103831-124 76718-626 76718-628 76718-630 77297-896 77312-414 77878-905
trans-4-(Boc-amino)cyclohexanemethylamine 95%
trans-4-(Boc-amino)cyclohexanemethylamine

Caution: for Research Use Only

Formula: C₁₂H₂₄N₂O₂
Boiling Pt: 342.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01861810
CAS Number: 177583-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR