Calcium glycerophosphate 95%

Supplier: Ambeed
Danger

A739815-10G
76697-654EA 8.49 USD
76697-654 77878-586
Calcium glycerophosphate 95%
Calcium glycerophosphate

Caution: for Research Use Only

Formula: C₃H₇CaO₆P
MDL Number: MFCD00042643
CAS Number: 27214-00-2
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR