4-Fluorophenyl isocyanate 97%

Supplier: Ambeed
Danger

Synonyms: 4-Fluorophenylisocyanate, 1-Fluoro-4-isocyanatobenzene

A655096-5G A655096-1G A655096-25G
76775-582EA 17 USD
76775-582 77573-102 77878-480
4-Fluorophenyl isocyanate 97%
4-Fluorophenyl isocyanate

Caution: for Research Use Only

Formula: C₇H₄FNO
MW: 137.11 g/mol
MDL Number: MFCD00002023
CAS Number: 1195-45-5
UN: 2478
ADR: 3,6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR