Methyl N-acetyl-α-D-glucosaminide ≥95%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-acetamido-2-deoxy-alpha-d-glucopyranoside, N-((2S,3R,4R,5S,6R)-4,5-Dihydroxy-6-(hydroxymethyl)-2-methoxytetrahydro-2H-pyran-3-yl)acetamide, Methyl 2-acetamido-2-deoxy-α-D-glucopyranoside

A329314-1G A329314-250MG A329314-5G
77626-626EA 135.96 USD
77626-626 77626-628 77878-035
Methyl N-acetyl-α-D-glucosaminide ≥95%
Methyl N-acetyl-α-D-glucosaminide

Caution: for Research Use Only

Formula: C₉H₁₇NO₆
MDL Number: MFCD00056076
CAS Number: 6082-04-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR