4-(4-((1-(4-((trans-3-(4-Cyano-3-methoxyphenoxy)-2,2,4,4-tetramethylcyclobutyl)carbamoyl)phenyl)piperidin-4-yl)methyl)piperazin-1-yl)-N-((S)-2,6-dioxopiperidin-3-yl)-2-fluorobenzamide 98%

Supplier: Ambeed
Warning

Synonyms: ARV-766

A2005058-25MG A2005058-50MG A2005058-100MG A2005058-250MG
77567-900EA 539 USD
77567-900 77625-994 77877-601 77877-602
4-(4-((1-(4-((trans-3-(4-Cyano-3-methoxyphenoxy)-2,2,4,4-tetramethylcyclobutyl)carbamoyl)phenyl)piperidin-4-yl)methyl)piperazin-1-yl)-N-((S)-2,6-dioxopiperidin-3-yl)-2-fluorobenzamide 98%
4-(4-((1-(4-((trans-3-(4-Cyano-3-methoxyphenoxy)-2,2,4,4-tetramethylcyclobutyl)carbamoyl)phenyl)piperidin-4-yl)methyl)piperazin-1-yl)-N-((S)-2,6-dioxopiperidin-3-yl)-2-fluorobenzamide

Caution: For research use only.

Formula: C₄₅H₅₄FN₇O₆
MW: 807.952 g/mol
CAS Number: 2750830-09-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR