3-Hydroxy-4-methoxycinnamic acid 98%

Supplier: Ambeed
Warning

Synonyms: Isoferulic acid, 3-(3-Hydroxy-4-methoxyphenyl)acrylic acid

A102256-10G A102256-25G A102256-25MG A102256-1G A102256-5G A102256-100G
103796-346EA 11 USD
103796-346 103822-038 76687-072 76687-074 76687-076 77308-916 77308-918 77876-790
3-Hydroxy-4-methoxycinnamic acid 98%
3-Hydroxy-4-methoxycinnamic acid

Caution: for Research Use Only

Formula: C₁₀H₁₀O₄
MW: 194.184 g/mol
Boiling Pt: 410.2 °C (760 mm Hg)
Melting Pt: 230 °C
Flash Pt: 167.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00004391
CAS Number: 537-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR