4-Propylphenol 98%

Supplier: Ambeed
Danger

Synonyms: p-Propylphenol, 4-n-Propylphenol

A1501778-25G A1501778-5G
76666-952EA 24 USD
76666-952 77875-751
4-Propylphenol 98%
4-Propylphenol

Caution: for Research Use Only

Formula: C₉H₁₂O
MDL Number: MFCD00002395
CAS Number: 645-56-7
UN: 2430
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR