Polyethylene terephthalate White granular, viscosity: 0.8-0.83mPa·s

Supplier: Ambeed
Warning

A1467715-100G
77875-049EA 17 USD
77875-049
Polyethylene terephthalate White granular, viscosity: 0.8-0.83mPa·s
Polyethylene terephthalate

Caution: for Research Use Only

Formula: (C₁₀H₈O₄)n
MDL Number: MFCD00084422
CAS Number: 25038-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR