Polyethylene glycol monostearate n≈40

Supplier: Ambeed
Warning

A1460312-100G
77874-962EA 11 USD
77874-962
Polyethylene glycol monostearate n≈40
Polyethylene glycol monostearate

Caution: for Research Use Only

Formula: HO(CH₂CH₂O)nOCC₁₇H₃₅
MDL Number: MFCD00148007
CAS Number: 9004-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR