N,N-Dimethyl-1,4-phenylenediammonium dichloride ≥96% AR, environmental protection reagent

Supplier: Ambeed
Danger

A1457480-5G
77874-950EA 9 USD
77874-950
N,N-Dimethyl-1,4-phenylenediammonium dichloride ≥96% AR, environmental protection reagent
N,N-Dimethyl-1,4-phenylenediammonium dichloride

Caution: for Research Use Only

Formula: (CH₃)₂NC₆H₄NH₂·2HCl
MDL Number: MFCD00012991
CAS Number: 536-46-9
UN: 2811
ADR: 6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR