Ethyl oleate 75+% (by GC)
Supplier: Ambeed
|
Warning
|
Synonyms:
Elaidic acid ethyl ester, (Z)-9-Octadecenoic acid ethyl ester, Oleic acid ethyl ester
A1453636-1000G
A1453636-25G
A1453636-100G
A1453636-500G
77622-162EA
41.2
USD
77622-162
77622-164
77874-924
77874-925
Ethyl oleate 75+% (by GC)
Ethyl oleate
Caution: for Research Use Only
Formula:
C₂₀H₃₈O₂ |
MDL Number:
MFCD00009579 CAS Number: 111-62-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...