Velocity™ Aluminum Lockout Padlocks, Brady
Supplier: Brady Worldwide
177281 177282 177283 177300 177301 177302 177303 177280
77873-788PK 389.99 USD
77873-788 77873-724 77873-748 77873-762 77873-712 77873-738 77873-696 77873-704
Velocity™ Aluminum Lockout Padlocks, Brady
Security Locks
Durable Velocity™ lockout padlocks are available in the widest variety of colors, materials, and styles, and are ideal for any size lockout program.
  • Service temperature: −15 to 250 °F
  • Body size: 1.5W×0.75D×1.8" H
Padlocks meet OSHA requirements to be durable, substantial, standardized, identifiable, and exclusively for safety. Color-matched keys and padlock bodies help workers find the right key for the right lock fast. Constructed using FDA-approved grease for safe and compliant use in food and beverage facilities. Energy-isolating design prevents electrical conductivity from the shackle to the key for added protection.
Key-retaining cylinder prevents key removal in the unlocked position, minimizing lost keys and the risk of unauthorized access. Key codes engraved on the key and lock body to simplify record-keeping and traceability. Aluminum padlocks with steel shackles are corrosion-resistant and ideal for general lockout applications where added durability or tamper resistance are needed.
Certifications: OSHA 1910.147.
Ordering information: Individual (each) padlock orders include 1 key and 1 padlock. Padlocks ordered in a pack of 6 include 6 keys and 6 padlocks.