(S)-(+)-4-Chlorodinaphthol[2,1-d:1',2'-f][1,3,2]dioxaphosphepin ≥95%
Supplier: Ambeed
|
Danger
|
Synonyms:
(S)-4-Chloro-dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin
A816330-100MG
A816330-250MG
A816330-1G
A816330-5G
77603-070EA
52.28
USD
77603-070
77603-072
77670-152
77873-280
(S)-(+)-4-Chlorodinaphthol[2,1-d:1',2'-f][1,3,2]dioxaphosphepin ≥95%
(S)-(+)-4-Chlorodinaphthol[2,1-d:1',2'-f][1,3,2]dioxaphosphepin
Caution: for Research Use Only
Formula:
C₂₀H₁₂ClO₂P |
MDL Number:
MFCD02093048 CAS Number: 137156-22-0 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...