m,m,m-Tritolyl phosphate 98%

Supplier: Ambeed
Danger

A780260-1G A780260-25G A780260-5G
76791-566EA 14 USD
76791-566 76791-568 77873-239
m,m,m-Tritolyl phosphate 98%
m,m,m-Tritolyl phosphate

Caution: for Research Use Only

Formula: C₂₁H₂₁O₄P
MDL Number: MFCD00041907
CAS Number: 563-04-2
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR