Silver neodecanoate ≥97%

Supplier: Ambeed
Warning

A655921-25G
77873-054EA 337 USD
77873-054
Silver neodecanoate ≥97%
Silver neodecanoate

Caution: for Research Use Only

Formula: C₁₀H₁₉AgO₂
MDL Number: MFCD01940647
CAS Number: 68683-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR