Trigonelline ≥95%

Supplier: Ambeed
Warning

Synonyms: 1-Methylpyridin-1-ium-3-carboxylate

A498292-250MG A498292-100MG A498292-1G
77602-146EA 227.55 USD
77602-146 77669-236 77872-832
Trigonelline ≥95%
Trigonelline

Caution: for Research Use Only

Formula: C₇H₇NO₂
MW: 137.15 g/mol
MDL Number: MFCD00054262
CAS Number: 535-83-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR