Suramin hexasodium 98%

Supplier: Ambeed
Warning

A397022-1MG A397022-100MG A397022-50MG A397022-1G A397022-250MG
76788-912EA 42 USD
76788-912 77668-826 77668-828 77872-622 77872-623
Suramin hexasodium 98%
Suramin hexasodium

Caution: for Research Use Only

Formula: C₅₁H₃₄N₆Na₆O₂₃S₆
MDL Number: MFCD00210217
CAS Number: 129-46-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR