Nα-Benzyloxycarbonyl-L-lysine thiobenzyl ester hydrochloride ≥97%
Supplier: Ambeed
|
Warning
|
A317143-1G
A317143-250MG
A317143-100MG
A317143-50MG
77668-402EA
571
USD
77668-402
77668-404
77872-445
77872-446
Nα-Benzyloxycarbonyl-L-lysine thiobenzyl ester hydrochloride ≥97%
Nα-Benzyloxycarbonyl-L-lysine thiobenzyl ester hydrochloride
Caution: for Research Use Only
Formula:
C₂₁H₂₇ClN₂O₃S |
MDL Number:
MFCD00058089 CAS Number: 69861-89-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...