trans-Vaccenic acid ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
trans-Octadec-11-enoic acid, Trans-11-Octadecenoic acid
A180388-100MG
A180388-250MG
A180388-1G
77667-490EA
210
USD
77667-490
77667-492
77871-902
trans-Vaccenic acid ≥98%
trans-Vaccenic acid
Caution: for Research Use Only
Formula:
C₁₈H₃₄O₂ MW: 282.468 g/mol |
MDL Number:
MFCD00002734 CAS Number: 693-72-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...