Butyl N-(3-Trifluoromethylphenyl)anthranilate 98%

Supplier: Ambeed
Warning

A149524-25G A149524-50MG A149524-5G A149524-100G A149524-1G
76783-312EA 34 USD
76783-312 76783-314 76783-316 77871-732 77871-733
Butyl N-(3-Trifluoromethylphenyl)anthranilate 98%
Butyl N-(3-Trifluoromethylphenyl)anthranilate

Caution: for Research Use Only

Formula: C₁₈H₁₈F₃NO₂
MDL Number: MFCD00866001
CAS Number: 67330-25-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR