Saturix 70% Presaturated Polyester Wipes, Validated Sterile, in Pouches
Supplier: FG CLEAN WIPES TE
6-LS7030VS-99P-N40
77871-048CS 762.19 USD
77871-048
Saturix 70% Presaturated Polyester Wipes, Validated Sterile, in Pouches
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Convenience, consistency, and cleanliness.
  • Validated sterile and lot traceable
  • 70% IPA, 30% DI water
  • Convenient resealable pouch
  • Versatile all-purpose wiping
  • Reduced VOCs with minimized bulk liquid storage and VOCs
  • Consistency in solvent application
Combining the benefits of validated sterility, a clean non-woven design, and the all-purpose concentration of 70% IPA, these wipes are versatile and dependable. Packaged in a convenient resealable pouch, a range of wet wiping tasks that require validated sterility become more cost effective and efficient.
Packaging:
Wipes are packaged 40 per pouch, 21 pouches per case.