Saturix Presaturated Cleanroom Wipes, 70% Presaturated Sachets
Supplier: FG CLEAN WIPES TE
6-B43
77871-042CS 343 USD
77871-042
Saturix Presaturated Cleanroom Wipes, 70% Presaturated Sachets
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Convenience, consistency, and cleanliness.
  • Reduces IPA use and minimizes bulk liquid storage
  • Reduces airborne VOC’s
  • Provides consistency in solvent application
  • Multiple sizes and packaging configurations in different formats and IPA concentrations
  • Variety of substrates for different critical environments
  • Easier and safer to ship, store, and use
This family of presaturated wipes feature wide-ranging substrates, IPA saturant levels, and packaging configurations for a multitude of environments and applications.
Packaging:
Wipes are packaged 100 per box, 10 boxes per case.