FARRAR® CYCLONE™ Ultra-Low Freeze/Store Chamber
Supplier: TRANE TECHNOLOGIES HOLDCO INC.
5828133-1
77871-034EA 28500 USD
77871-034
FARRAR® CYCLONE™ Ultra-Low Freeze/Store Chamber
Freezers Ultra Low Temperature Freezers
A purpose built freeze/store chamber for pre-clinical to production scale needs. The CYCLONE uses forced-air convection to help ensure powerful cooling performance with chest freezer-like temperature recovery with the convenience of an upright.
  • Flexible temperature setpoint (−20 to −80 °C) and tight uniformity (±1 °C at −80 °C and ±2 °C at −20 °C)
  • Purpose built for freezing and storage operations with 3× freezing capacity over standard ULTs
  • Fast temperature recovery enables frequent access, minimizes excursions from door openings, and helps protect sample integrity
  • Sustainability and performance with low GWP, hydrocarbon refrigerants and ENERGY STAR® compliance
  • Maximized volume-to-footprint ratio Modular refrigeration system enables quick swap with minimal operation impact
  • Integrated communications with BMS/BAS systems and monitoring programs via Modbus TCP/IP and BACnet IP
Blending industry-leading, ENERGY STAR® compliant sustainability with unmatched cooling performance, FARRAR CYCLONE provides pull-down, door-opening recovery, and temperature uniformity unheard of in conventional cold wall freezers, traditional ultra-low temperature freezers, and standard cold storage solutions. A packaged, quick-change top-mounted refrigeration system ensures easy, point-of-use maintenance. Heat rejection on top of the unit helps eliminate footprint clearance requirements, increasing density without impacting performance.
Delivery information: Supplied with three shelves. Additional shelves are available as optional accessories.