5-Methyl-Cytidine-5-Triphosphate (5-Methyl-CTP)
Supplier: New England Biolabs (NEB)
N0432S
77868-090EA 758.5 USD
77868-090
5-Methyl-Cytidine-5-Triphosphate (5-Methyl-CTP)
Nucleic Acid Reagents Nucleotides
mRNA has emerged as a potent modality for therapeutics.
  • Supplied as sodium salts, pH 7.0
  • Modified NTPs are commonly used for reduction of immunogenicity for in vitro transcription RNA
  • No detectable RNase, DNase, phosphatase or protease contaminating activities
  • ≥95% purity, as determined by HPLC
  • Optimal activity and stability for up to 24 months
  • Concentration: 100 mM
Incorporation of chemically modified ribonucleotides improves the efficacy of these therapies by increasing stability, reducing immunogenicity, and enhancing translatability of mRNAs.