Polyethylene glycol monomethyl ether, M.W. ~1000
Supplier: Ambeed
|
Warning
|
Synonyms:
Methoxypolyethylene glycols, Methoxy poly(ethylene glycol), PEG MME, Poly(ethylene glycol) methyl ether, mPEG, Methoxypolyethylene glycols-average M.W. 1000, m-PEG-OH
A1460296-100G
A1460296-25G
A1460296-1000G
A1460296-500G
77597-292EA
10.25
USD
77597-292
77597-294
77622-192
77867-818
Polyethylene glycol monomethyl ether, M.W. ~1000
Polyethylene glycol monomethyl ether
Caution: for Research Use Only
Formula:
CH₃(OCH₂CH₂)nOH |
MDL Number:
MFCD00084416 CAS Number: 9004-74-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...