GDC-046 ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,6-Dichloro-N-(2-(cyclopropanecarboxamido)pyridin-4-yl)benzamide
A1325556-100MG
A1325556-250MG
77867-278EA
231
USD
77867-278
77867-279
GDC-046 ≥98%
GDC-046
Caution: for Research Use Only
Formula:
C₁₆H₁₃Cl₂N₃O₂ |
MDL Number:
MFCD28155087 CAS Number: 1258292-64-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...