NexION® 2200 ICP Mass Spectrometer
Supplier: PerkinElmer
N8170012
77865-778EA 261100 USD
77865-778
NexION® 2200 ICP Mass Spectrometer
Spectrophotometers and Photometers Mass Spectrometers
For analytical testing labs performing trace-elemental analyses, the NexION® 2200 ICP-MS builds on a 40-year history of innovation in ICP-MS and a strong tradition of reliable, easy-to-use, low-maintenance instrumentation, delivering uncompromised performance, accuracy, and repeatability.
  • Reliable
  • Easy-to-use
Its unique three-quad design, combined with novel and proven technologies, including the second-generation Triple Cone Interface with OmniRing™, come together to offer outstanding sensitivity, superior interference removal, excellent stability, unmatched matrix tolerance, high throughput/greater uptime, as well as sustainability and low cost of operation.
Weight: 150.0 kg