NexION® 1100 ICP Mass Spectrometer
Supplier: PerkinElmer
N8170011
77865-776EA 216713 USD
77865-776
NexION® 1100 ICP Mass Spectrometer
Spectrophotometers and Photometers Mass Spectrometers
For testing labs conducting trace-elemental analyses looking for a reliable yet easy-to operate system capable of high sample throughput, the NexION® 1100 ICP-MS builds on a strong tradition of reliable, easy-to-use, low-maintenance instrumentation, now reinventing the user experience for routine ICP-MS applications.
  • Reliable
  • Easy-to-use
Its unique single-analyzer, three-quad design, combined with a single gas channel (with upgrade option to multiple channels) and a modernized workflow, come together to offer simple yet effective interference removal, low maintenance, excellent stability, as well as sustainability, all in a streamlined experience - the ideal ICP-MS for day-to-day trace-elemental analyses.
Weight: 150.0 kg