PinAAcle™ 500 Flame Atomic Absorption Spectrometer
Supplier: PerkinElmer
N3180011
77865-770EA 35100 USD
77865-770
PinAAcle™ 500 Flame Atomic Absorption Spectrometer
Spectrophotometers and Photometers Mass Spectrometers
Our PinAAcle™ 500 is the world’s first completely corrosion-resistant flame atomic absorption (AA) spectrometer, designed to withstand the harshest environments and most corrosive samples. It offers superior durability, longer life, lower maintenance costs, and the fastest return on investment of any flame AA.
  • Superior durability
A robust, fully-integrated, flame-only benchtop system, ideal for laboratories needing a reliable, easy-to-use, yet high-performance flame AA. It is equipped with an innovative touchscreen interface with the flexibility to operate via its easy-to-use Syngistix Touch™ software or the more comprehensive, optional Syngistix™ for AA Software.
Weight: 57.0 kg