Avio® 550 Max ICP-OES Configuration
Supplier: PerkinElmer
N0810005
77865-768EA 124200 USD
77865-768
Avio® 550 Max ICP-OES Configuration
Spectrophotometers and Photometers Mass Spectrometers
The Avio® 550 Max is a compact, fully simultaneous ICP-OES instrument, ideal for labs with high throughput requirements.
  • Productivity, performance and faster return on investment
It utilizes a vertical plasma and is engineered to handle even the most difficult, high-matrix samples without dilution, delivering productivity, performance and faster return on investment. Plus, Syngistix™ for ICP software provides an intuitive and smart environment with smart monitoring for real-time instrument and sample diagnostics as well as results viewing to easily track sample analysis, quality control, and internal standard performance to guarantee sample accuracy.
Weight: 163.0 kg