Avio® 220 Max ICP-OES Configuration
Supplier: PerkinElmer
N0790005
77865-766EA 97000 USD
77865-766
Avio® 220 Max ICP-OES Configuration
Spectrophotometers and Photometers Mass Spectrometers
The Avio® 220 Max is a compact, hybrid simultaneous ICP-OES instrument, ideal for labs with low-to-medium throughput requirements.
  • Fast startup
Its unique plug-and-play capabilities make the Avio® 220 Max the ICP with the fastest startup on the market - cold start to analysis in only 10 minutes - saving you time and operating costs by allowing you to shut down the instrument between runs. Plus, it utilizes a vertical plasma and is engineered to handle even the most difficult, high-matrix samples without dilution, delivering productivity, performance and faster return on investment.
And last but certainly not least, Syngistix™ for ICP software provides an intuitive and smart environment with smart monitoring for real-time instrument and sample diagnostics as well as results viewing to easily track sample analysis, quality control, and internal standard performance to guarantee sample accuracy.
Weight: 100.0 kg