Dr. Ehrenstorfer Perfluorotridecanoic Acid 13C2 (1,2-13C2) 50 µg/ml in Methanol:Water
Supplier: LGC Standards
DRE-A15988003MW-50
77864-020EA 518 USD
77864-020
Dr. Ehrenstorfer Perfluorotridecanoic Acid 13C2 (1,2-13C2) 50 µg/ml in Methanol:Water
Standards Environmental Analysis Standards