Sucrose (60.00 °Brix / 1.44193 RI), Sucrose Brix/RI Standard, LGC Industrial
Supplier: LGC Standards
ALK-SS60
77863-226EA
69
USD
77863-226
Sucrose (60.00 °Brix / 1.44193 RI), Sucrose Brix/RI Standard, LGC Industrial
Standards
Refractive Index Standards