Sucrose (35.00 °Brix / 1.39032 RI), Sucrose Brix/RI Standard, LGC Industrial
Supplier: LGC Standards
ALK-SS35
77863-221EA
69
USD
77863-221
Sucrose (35.00 °Brix / 1.39032 RI), Sucrose Brix/RI Standard, LGC Industrial
Standards
Refractive Index Standards