Sucrose (30.00 °Brix / 1.38115 RI), Sucrose Brix/RI Standard, LGC Industrial
Supplier: LGC Standards
ALK-SS30
77863-220EA
69
USD
77863-220
Sucrose (30.00 °Brix / 1.38115 RI), Sucrose Brix/RI Standard, LGC Industrial
Standards
Refractive Index Standards