Sucrose (20.00 °Brix / 1.36384 RI), Sucrose Brix/RI Standard, LGC Industrial
Supplier: LGC Standards
ALK-SS20
77863-218EA
69
USD
77863-218
Sucrose (20.00 °Brix / 1.36384 RI), Sucrose Brix/RI Standard, LGC Industrial
Standards
Refractive Index Standards