Dispersion Blade Set
Supplier: Caframo
A400SET
77860-238EA 614.33 USD
77860-238
Dispersion Blade Set
Stirrers Stirring Tools
Dispersion Blade Set with 8 mm dia (⁵⁄₁₆") bore - includes three different blades for high shear mixing at medium to high speeds.
  • Electropolished 316 stainless steel
  • High shear
  • Quick changes to test and optimize different batch sizes
  • Chemically inert and corrosion resistant
  • Broken particles are efficiently dispersed
Ordering information:
Shaft available separately.
Delivery information:
Set includes 48 mm (1⅞") A164, 75 mm (3") A431, and 100 mm (4") A441 blades with set screws. Blades are composed of electropolished 316 stainless steel with an 8 mm (⁵⁄₁₆") bore.