Anti-LCT Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A27369-100UG A27369-50UG
77742-037EA 330 USD
77742-037 77858-875
Anti-LCT Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: LCT
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR